3-Amino-2-(benzyloxy)-5-chlorophenylbenzenecarboxylate 95+%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-2-(benzyloxy)-5-chlorophenyl benzoate

OR40056-500MG
APOSOR40056-500MGEA 661 EUR
APOSOR40056-500MG
3-Amino-2-(benzyloxy)-5-chlorophenylbenzenecarboxylate 95+%
3-Amino-2-(benzyloxy)-5-chlorophenylbenzenecarboxylate
Formel: C₂₀H₁₆ClNO₃
Molecular Weight: 353.81 g/mol
Schmelzpunkt: 140…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16140258
CAS-Nummer: 1221792-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR