3-(Dimethylamino)benzoesäure

Lieferant: Apollo Scientific

OR400573-100G OR400573-500G
APOSOR400573-100GEA 0 EUR
APOSOR400573-100G APOSOR400573-500G
3-(Dimethylamino)benzoesäure
3-(Dimethylamino)benzoesäure
Formel: C₉H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00002497
CAS-Nummer: 99-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR