5-Bromo-2-iodo-4-methoxybenzaldehyde 95+%
Lieferant: Apollo Scientific
OR400578-250MG
APOSOR400578-250MGEA
130
EUR
APOSOR400578-250MG
5-Bromo-2-iodo-4-methoxybenzaldehyde 95+%
5-Bromo-2-iodo-4-methoxybenzaldehyde
Formel:
C₈H₆BrIO₂ Molecular Weight: 340,941 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17014846 CAS-Nummer: 1269293-42-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...