4-Methylnicotinoyl chloride

Lieferant: Apollo Scientific

OR40060-1G OR40060-250MG OR40060-500MG OR40060-2.5G
APOSOR40060-1GEA 336 EUR
APOSOR40060-1G APOSOR40060-250MG APOSOR40060-500MG APOSOR40060-2.5G
4-Methylnicotinoyl chloride
4-Methylnicotinoyl chloride
Formel: C₇H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD08751335
CAS-Nummer: 155136-54-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR