5-Bromo-2-nitrobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

OR400606-1G OR400606-250MG OR400606-5G
APOSOR400606-1GEA 0 EUR
APOSOR400606-1G APOSOR400606-250MG APOSOR400606-5G
5-Bromo-2-nitrobenzyl bromide
5-Bromo-2-nitrobenzyl bromide
Formel: C₇H₅Br₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11848689
CAS-Nummer: 35287-42-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR