2-(Dimethylamino)benzoesäure 97%

Lieferant: Apollo Scientific

Synonyme: 2-Dimethylaminobenzoic acid

OR400613-5G
APOSOR400613-5GEA 71.2 EUR
APOSOR400613-5G
2-(Dimethylamino)benzoesäure 97%
2-(Dimethylamino)benzoesäure
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016496
CAS-Nummer: 610-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR