Methyl-5-amino-2-methoxybenzoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 5-amino-2-methoxybenzoate
OR40062-1G
APOSOR40062-1GEA
78.3
EUR
APOSOR40062-1G
Methyl-5-amino-2-methoxybenzoate
Methyl-5-amino-2-methoxybenzoate
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08458835 CAS-Nummer: 22802-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...