4-(Dimethylamino)benzyl bromide hydrobromide, tech

Lieferant: Apollo Scientific
Gefahr

OR400627-1G OR400627-250MG
APOSOR400627-1GEA 522 EUR
APOSOR400627-1G APOSOR400627-250MG
4-(Dimethylamino)benzyl bromide hydrobromide, tech
4-(Dimethylamino)benzyl bromide hydrobromide
Formel: C₉H₁₂BrN·BrH
Lagertemperatur: Raumtemperatur
MDL: MFCD13195837
CAS-Nummer: 1116572-69-0
UN: 2928
ADR: 6.1+8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR