Methyl 4-(Bromomethyl)-3-iodobenzoate 95%

Lieferant: Apollo Scientific
Achtung

OR400640-1G
APOSOR400640-1GEA 509 EUR
APOSOR400640-1G
Methyl 4-(Bromomethyl)-3-iodobenzoate 95%
Methyl 4-(Bromomethyl)-3-iodobenzoate
Formel: C₉H₈BrIO₂
Molecular Weight: 354,97 g/mol
MDL: MFCD13659352
CAS-Nummer: 229028-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR