3-Chlor-2-hydroxybenzonitril

Lieferant: Apollo Scientific
Achtung

OR400649-1G OR400649-25G OR400649-5G
APOSOR400649-1GEA 24.8 EUR
APOSOR400649-1G APOSOR400649-25G APOSOR400649-5G
3-Chlor-2-hydroxybenzonitril
3-Chlor-2-hydroxybenzonitril
Formel: C₇H₄ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09999455
CAS-Nummer: 13073-27-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR