3-Chlor-2-hydroxybenzonitril
Lieferant: Apollo Scientific
|
Achtung
|
OR400649-1G
OR400649-25G
OR400649-5G
APOSOR400649-1GEA
24.8
EUR
APOSOR400649-1G
APOSOR400649-25G
APOSOR400649-5G
3-Chlor-2-hydroxybenzonitril
3-Chlor-2-hydroxybenzonitril
Formel:
C₇H₄ClNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09999455 CAS-Nummer: 13073-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...