1-Bromo-3-chloro-2-iodobenzene 99%

Lieferant: Apollo Scientific
Achtung

OR400660-25G OR400660-5G
APOSOR400660-25GEA 145 EUR
APOSOR400660-25G APOSOR400660-5G
1-Bromo-3-chloro-2-iodobenzene 99%
1-Bromo-3-chloro-2-iodobenzene
Formel: C₆H₃BrClI
Lagertemperatur: Raumtemperatur
MDL: MFCD09907935
CAS-Nummer: 450412-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR