6-Methylpyrazine-2-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR40068-1G
APOSOR40068-1GEA 29 EUR
APOSOR40068-1G
6-Methylpyrazine-2-carboxylic acid 97%
6-Methylpyrazine-2-carboxylic acid
Formel: C₆H₆N₂O₂
Molecular Weight: 138,1200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705756
CAS-Nummer: 5521-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR