Methyl 2-bromo-4-hydroxybenzoate 95%

Lieferant: Apollo Scientific
Achtung

OR400687-1G
APOSOR400687-1GEA 323 EUR
APOSOR400687-1G
Methyl 2-bromo-4-hydroxybenzoate 95%
Methyl 2-bromo-4-hydroxybenzoate
Formel: C₈H₇BrO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18398783
CAS-Nummer: 101085-03-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR