6-Methylpyrazin-2-ol 98%

Lieferant: Apollo Scientific

Synonyme: 2-Hydroxy-6-methylpyrazine

OR40069-25G
APOSOR40069-25GEA 143 EUR
APOSOR40069-25G
6-Methylpyrazin-2-ol 98%
6-Methylpyrazin-2-ol
Formel: C₅H₆N₂O
Molecular Weight: 110,1139 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705761
CAS-Nummer: 20721-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR