4-Bromo-2-iodo-6-nitroanisole

Lieferant: Apollo Scientific
Achtung

OR400691-1G OR400691-5G
APOSOR400691-1GEA 183 EUR
APOSOR400691-1G APOSOR400691-5G
4-Bromo-2-iodo-6-nitroanisole
4-Bromo-2-iodo-6-nitroanisole
Formel: C₇H₅BrINO₃
Molecular Weight: 357.928 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28898061
CAS-Nummer: 1935459-09-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR