4-Bromo-2-iodo-6-nitroanisole
Lieferant: Apollo Scientific
|
Achtung
|
OR400691-1G
OR400691-5G
APOSOR400691-1GEA
183
EUR
APOSOR400691-1G
APOSOR400691-5G
4-Bromo-2-iodo-6-nitroanisole
4-Bromo-2-iodo-6-nitroanisole
Formel:
C₇H₅BrINO₃ Molecular Weight: 357.928 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28898061 CAS-Nummer: 1935459-09-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...