5-Bromo-2-chlorophenacyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

OR400708-250MG OR400708-1G OR400708-5G
APOSOR400708-250MGEA 0 EUR
APOSOR400708-250MG APOSOR400708-1G APOSOR400708-5G
5-Bromo-2-chlorophenacyl bromide 97%
5-Bromo-2-chlorophenacyl bromide
Formel: C₈H₅Br₂ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD18392520
CAS-Nummer: 1261775-99-8
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR