2-Bromo-3-iodobenzonitrile

Lieferant: Apollo Scientific
Achtung

OR400744-1G OR400744-250MG
APOSOR400744-1GEA 574 EUR
APOSOR400744-1G APOSOR400744-250MG
2-Bromo-3-iodobenzonitrile
2-Bromo-3-iodobenzonitrile
Formel: C₇H₃BrIN
Molecular Weight: 307.914 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391642
CAS-Nummer: 1261498-04-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR