Methyl 2-bromo-5-formylbenzoate

Lieferant: Apollo Scientific
Achtung

OR400745-250MG
APOSOR400745-250MGEA 52.2 EUR
APOSOR400745-250MG
Methyl 2-bromo-5-formylbenzoate
Methyl 2-bromo-5-formylbenzoate
Formel: C₉H₇BrO₃
Molecular Weight: 243,054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26863463
CAS-Nummer: 1043418-95-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR