2-Brom-6-iodbenzonitril
Lieferant: Apollo Scientific
OR400749-5G
APOSOR400749-5GEA
165
EUR
APOSOR400749-5G
2-Brom-6-iodbenzonitril
2-Brom-6-iodbenzonitril
Formel:
C₇H₃BrIN Molecular Weight: 307,9139 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16619725 CAS-Nummer: 1245648-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...