5-amino-2-bromo-4-chloroanisole 98%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-chloro-5-methoxyaniline

OR400852-1G
APOSOR400852-1GEA 120 EUR
APOSOR400852-1G
5-amino-2-bromo-4-chloroanisole 98%
5-amino-2-bromo-4-chloroanisole
Formel: C₇H₇BrClNO
Molecular Weight: 236,494 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845922
CAS-Nummer: 98446-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR