8-Brom-2,4-dichlorchinazolin 98%

Lieferant: Apollo Scientific

OR400858-1G
APOSOR400858-1GEA 116 EUR
APOSOR400858-1G
8-Brom-2,4-dichlorchinazolin 98%
8-Bromo-2,4-dichloroquinazoline
Formel: C₈H₃BrCl₂N₂
Molecular Weight: 277,93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954891
CAS-Nummer: 331647-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR