1-Boc-3-(bromomethyl)piperidine

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-3-(bromomethyl)piperidine, 3-(Bromomethyl)piperidine, N-BOC protected

OR40087-100MG
APOSOR40087-100MGEA 21.9 EUR
APOSOR40087-100MG
1-Boc-3-(bromomethyl)piperidine
1-Boc-3-(bromomethyl)piperidine
Formel: C₁₁H₂₀BrNO₂
Molecular Weight: 278,1900 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD04115539
CAS-Nummer: 193629-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR