4-Bromo-3-methoxybenzenesulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR400875-1G OR400875-250MG OR400875-5G
APOSOR400875-1GEA 48 EUR
APOSOR400875-1G APOSOR400875-250MG APOSOR400875-5G
4-Bromo-3-methoxybenzenesulphonyl chloride 95%
4-Bromo-3-methoxybenzenesulphonyl chloride
Formel: C₇H₆BrClO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD14702923
CAS-Nummer: 1215295-91-2
UN: 1759
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR