6-Bromo-3-iodo-2-nitrophenol

Lieferant: Apollo Scientific
Achtung

OR400888-250MG OR400888-1G
APOSOR400888-250MGEA 174 EUR
APOSOR400888-250MG APOSOR400888-1G
6-Bromo-3-iodo-2-nitrophenol
6-Bromo-3-iodo-2-nitrophenol
Formel: C₆H₃BrINO₃
Molecular Weight: 343.901 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD30730276
CAS-Nummer: 2090513-56-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR