5-Acetylthiophene-2-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Acetyl-5-formylthiophene, 5-Acetyl-2-thenaldehyde, 5-Acetylthiophene-2-carbaldehyde, 2-Ethanoyl-5-formylthiophene

OR4009-100MG
APOSOR4009-100MGEA 21.9 EUR
APOSOR4009-100MG
5-Acetylthiophene-2-carboxaldehyde
5-Acetylthiophene-2-carboxaldehyde
Formel: C₇H₆O₂S
Molecular Weight: 154.186 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00509280
CAS-Nummer: 4565-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR