4-Bromo-2-(methylsulphonyl)phenol 95+%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-methanesulphonylphenol

OR400916-250MG
APOSOR400916-250MGEA 125 EUR
APOSOR400916-250MG
4-Bromo-2-(methylsulphonyl)phenol 95+%
4-Bromo-2-(methylsulphonyl)phenol
Formel: C₇H₇BrO₃S
Molecular Weight: 251.098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28515417
CAS-Nummer: 88041-67-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR