2-Bromo-6-chlorophenacyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

OR400928-1G OR400928-250MG
APOSOR400928-1GEA 0 EUR
APOSOR400928-1G APOSOR400928-250MG
2-Bromo-6-chlorophenacyl bromide 95%
2-Bromo-6-chlorophenacyl bromide
Formel: C₈H₅Br₂ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD18392521
CAS-Nummer: 1261604-22-1
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR