Benzo[b]thiophene-4-carboxylic acid

Lieferant: Apollo Scientific

OR400932-1G
APOSOR400932-1GEA 132 EUR
APOSOR400932-1G
Benzo[b]thiophene-4-carboxylic acid
Benzo[b]thiophene-4-carboxylic acid
Formel: C₉H₆O₂S
Molecular Weight: 178.208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01929340
CAS-Nummer: 10134-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR