N-Methoxy-N,1-dimethylpiperidine-4-carboxamide 95+%

Lieferant: Apollo Scientific

OR400941-1G OR400941-5G
APOSOR400941-1GEA 42.2 EUR
APOSOR400941-1G APOSOR400941-5G
N-Methoxy-N,1-dimethylpiperidine-4-carboxamide 95+%
N-Methoxy-N,1-dimethylpiperidine-4-carboxamide
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186.251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22575146
CAS-Nummer: 215950-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR