5-Benzo[b]thiophencarbonsäure

Lieferant: Apollo Scientific

OR400942-1G
APOSOR400942-1GEA 0 EUR
APOSOR400942-1G
5-Benzo[b]thiophencarbonsäure
5-Benzo[b]thiophencarbonsäure
Formel: C₉H₆O₂S
Molecular Weight: 178.2077 g/mol
Schmelzpunkt: 211…215 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04974042
CAS-Nummer: 2060-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR