Benzo[b]thiophen-5-methanol

Lieferant: Apollo Scientific

OR400946-250MG
APOSOR400946-250MGEA 219 EUR
APOSOR400946-250MG
Benzo[b]thiophen-5-methanol
Benzo[b]thiophen-5-methanol
Formel: C₉H₈OS
Lagertemperatur: Raumtemperatur
CAS-Nummer: 20532-34-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR