2-Bromo-4-iodobenzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Bromo-4-iodophenyl)methanol

OR400950-1G
APOSOR400950-1GEA 52.2 EUR
APOSOR400950-1G
2-Bromo-4-iodobenzyl alcohol
2-Bromo-4-iodobenzyl alcohol
Formel: C₇H₆BrIO
Molecular Weight: 312.93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11520411
CAS-Nummer: 1261648-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR