2,2'-Furil dioxime 97%

Lieferant: Apollo Scientific
Achtung

OR400953-5G OR400953-1G
APOSOR400953-5GEA 543 EUR
APOSOR400953-5G APOSOR400953-1G
2,2'-Furil dioxime 97%
2,2'-Furil dioxime

Mixture of isomers

Formel: C₁₀H₈N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00075315
CAS-Nummer: 522-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR