6-Bromo-1H-indazole-3-carboxylic acid 96%

Lieferant: Apollo Scientific
Achtung

OR40096-100MG
APOSOR40096-100MGEA 26.1 EUR
APOSOR40096-100MG
6-Bromo-1H-indazole-3-carboxylic acid 96%
6-Bromo-1H-indazole-3-carboxylic acid
Formel: C₈H₅BrN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD05663977
CAS-Nummer: 660823-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR