2-Chlor-5-methoxybenzaldehyd

Lieferant: Apollo Scientific

OR400965-5G
APOSOR400965-5GEA 114 EUR
APOSOR400965-5G
2-Chlor-5-methoxybenzaldehyd
2-Chloro-5-methoxybenzaldehyde
Formel: C₈H₇ClO₂
Molecular Weight: 170.5900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11845925
CAS-Nummer: 13719-61-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR