2-Chloro-6-iodobenzoic acid

Lieferant: Apollo Scientific

OR400966-5G
APOSOR400966-5GEA 56.7 EUR
APOSOR400966-5G
2-Chloro-6-iodobenzoic acid
2-Chloro-6-iodobenzoic acid
Formel: C₇H₄ClIO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06808823
CAS-Nummer: 13420-63-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR