3-Chloro-4-iodobenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR400981-1G
OR400981-250MG
OR400981-5G
APOSOR400981-1GEA
0
EUR
APOSOR400981-1G
APOSOR400981-250MG
APOSOR400981-5G
3-Chloro-4-iodobenzyl bromide 95%
3-Chloro-4-iodobenzyl bromide
Formel:
C₇H₅BrClI Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393368 CAS-Nummer: 166386-59-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...