3-Chloro-4-iodobenzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

OR400981-1G OR400981-250MG OR400981-5G
APOSOR400981-1GEA 0 EUR
APOSOR400981-1G APOSOR400981-250MG APOSOR400981-5G
3-Chloro-4-iodobenzyl bromide 95%
3-Chloro-4-iodobenzyl bromide
Formel: C₇H₅BrClI
Lagertemperatur: Raumtemperatur
MDL: MFCD18393368
CAS-Nummer: 166386-59-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR