3-Chloro-2-nitrobenzonitrile

Lieferant: Apollo Scientific

OR401015-5G
APOSOR401015-5GEA 133 EUR
APOSOR401015-5G
3-Chloro-2-nitrobenzonitrile
3-Chloro-2-nitrobenzonitrile
Formel: C₇H₃ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD15529744
CAS-Nummer: 34662-28-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR