3-Chloro-2-nitrobenzaldehyde

Lieferant: Apollo Scientific

OR401017-1G
APOSOR401017-1GEA 143 EUR
APOSOR401017-1G
3-Chloro-2-nitrobenzaldehyde
3-Chloro-2-nitrobenzaldehyde
Formel: C₇H₄ClNO₃
Molecular Weight: 185.565 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797953
CAS-Nummer: 22233-52-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR