3-Ethoxy-4-methoxyphenylacetylene

Lieferant: Apollo Scientific
Achtung

OR40102-1G
APOSOR40102-1GEA 393 EUR
APOSOR40102-1G
3-Ethoxy-4-methoxyphenylacetylene
3-Ethoxy-4-methoxyphenylacetylene
Formel: C₁₁H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11869745
CAS-Nummer: 881919-82-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR