4-Chlor-3-nitrobenzylalkohol

Lieferant: Apollo Scientific

OR401027-25G
APOSOR401027-25GEA 69.6 EUR
APOSOR401027-25G
4-Chlor-3-nitrobenzylalkohol
4-Chlor-3-nitrobenzylalkohol
Formel: C₇H₆ClNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00007086
CAS-Nummer: 55912-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR