3-Chloro-5-nitrobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

OR401031-1G OR401031-250MG
APOSOR401031-1GEA 0 EUR
APOSOR401031-1G APOSOR401031-250MG
3-Chloro-5-nitrobenzyl bromide
3-Chloro-5-nitrobenzyl bromide
Formel: C₇H₅BrClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11505896
CAS-Nummer: 865624-17-5
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR