2-Iodo-3-nitrobenzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

OR401054-1G OR401054-250MG OR401054-5G
APOSOR401054-1GEA 0 EUR
APOSOR401054-1G APOSOR401054-250MG APOSOR401054-5G
2-Iodo-3-nitrobenzyl bromide 95%
2-Iodo-3-nitrobenzyl bromide
Formel: C₇H₅BrINO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18398002
CAS-Nummer: 1261777-67-6
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR