3-Iodo-5-nitrobenzonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR401059-1G
OR401059-250MG
OR401059-5G
APOSOR401059-1GEA
522
EUR
APOSOR401059-1G
APOSOR401059-250MG
APOSOR401059-5G
3-Iodo-5-nitrobenzonitrile 95%
3-Iodo-5-nitrobenzonitrile
Formel:
C₇H₃IN₂O₂ Molecular Weight: 274.015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00464943 CAS-Nummer: 49674-30-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...