4-Hydroxy-3-(methoxycarbonyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methyl 4-hydroxyisophthalate, Methyl 5-carboxy-2-hydroxybenzoate

OR40106-1G OR40106-250MG
APOSOR40106-1GEA 567 EUR
APOSOR40106-1G APOSOR40106-250MG
4-Hydroxy-3-(methoxycarbonyl)benzoic acid 95%
4-Hydroxy-3-(methoxycarbonyl)benzoic acid
Formel: C₉H₈O₅
Molecular Weight: 196.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11869747
CAS-Nummer: 41684-11-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR