4-Acetyl-2-methylbenzoic acid
Lieferant: Apollo Scientific
OR401070-1G
APOSOR401070-1GEA
288
EUR
APOSOR401070-1G
4-Acetyl-2-methylbenzoic acid
4-Acetyl-2-methylbenzoic acid
Formel:
C₁₀H₁₀O₃ Molecular Weight: 178.185 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20639525 CAS-Nummer: 55860-35-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...