6-Iodoisoquinoline

Lieferant: Apollo Scientific
Achtung

OR40112-1G OR40112-250MG
APOSOR40112-1GEA 562 EUR
APOSOR40112-1G APOSOR40112-250MG
6-Iodoisoquinoline
6-Iodoisoquinoline
Formel: C₉H₆IN
Lagertemperatur: Raumtemperatur
MDL: MFCD11109663
CAS-Nummer: 75476-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR